7-Bromo-2-chloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2478G1 AS2478G5
ACTIAS2478G1EA 82 GBP
ACTIAS2478G1 ACTIAS2478G5
7-Bromo-2-chloroquinazoline
7-Bromo-2-chloroquinazoline
Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD11040175
CAS Number: 953039-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR