4-(Chloromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2333-5G BB-2333-1G
COBBBB-2333-5GEA 0 GBP
COBBBB-2333-5G COBBBB-2333-1G
4-(Chloromethyl)phenylboronic acid
4-(Chloromethyl)phenylboronic acid
Formula: C₇H₈BClO₂
MDL Number: MFCD07363850
CAS Number: 164413-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR