1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-(trifluoromethoxy)phenyl sulphone

OT-0649-5G OT-0649-25G OT-0649-1G
COBBOT-0649-5GEA 0 GBP
COBBOT-0649-5G COBBOT-0649-25G COBBOT-0649-1G
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
Formula: C₈H₇F₃O₃S
MDL Number: MFCD00792426
CAS Number: 87750-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR