Methyl-6-aminopicolinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Aminopicolinic acid methyll ester, Methyl 6-aminopicolinate, Methyl 6-Aminopicolinate, Methyl 6-Aminopyridine-2-carboxylate, 6-Aminopyridine-2-carboxylic acid methyll ester

AS32531G1 AS32531G5
ACTIAS32531G1EA 0 GBP
ACTIAS32531G1 ACTIAS32531G5
Methyl-6-aminopicolinate
Methyl 6-aminopicolinate
Formula: C₇H₈N₂O₂
MW: 152.1500 g/mol
MDL Number: MFCD00233712
CAS Number: 36052-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR