3-(Aminomethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Cyanobenzylamine

AS22574G1 AS22574G5
ACTIAS22574G1EA 71 GBP
ACTIAS22574G1 ACTIAS22574G5
3-(Aminomethyl)benzonitrile
3-(Aminomethyl)benzonitrile
Formula: C₈H₈N₂
MW: 132.1600 g/mol
MDL Number: MFCD06797832
CAS Number: 10406-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR