Ethyl-6-fluoro-4-hydroxyquinoline-3-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate

AS20840G1 AS20840G25 AS20840G5
ACTIAS20840G1EA 30 GBP
ACTIAS20840G1 ACTIAS20840G25 ACTIAS20840G5
Ethyl-6-fluoro-4-hydroxyquinoline-3-carboxylate
Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate
Formula: C₁₂H₁₀FNO₃
MW: 235.22 g/mol
MDL Number: MFCD00173345
CAS Number: 318-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR