Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 1,4-dihydro-8-fluoro-4-oxoquinoline-3-carboxylate

AS20757G25 AS20757G5
ACTIAS20757G25EA 0 GBP
ACTIAS20757G25 ACTIAS20757G5
Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate
Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate
Formula: C₁₂H₁₀FNO₃
MW: 235.211 g/mol
MDL Number: MFCD00052242
CAS Number: 63010-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR