3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008252-1G 008252-5G
FLUO008252-1GEA 0 GBP
FLUO008252-1G FLUO008252-5G
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Formula: C₁₀H₆ClF₃N₂O
MDL Number: MFCD00084969
CAS Number: 175205-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR