4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008232-1G 008232-5G
FLUO008232-1GEA 0 GBP
FLUO008232-1G FLUO008232-5G
4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde
4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde
Formula: C₁₄H₁₀ClFO₂
MDL Number: MFCD00052227
CAS Number: 172932-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR