4-(Isopropylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid

BB-3009-5G BB-3009-1G
COBBBB-3009-5GEA 0 GBP
COBBBB-3009-5G COBBBB-3009-1G
4-(Isopropylcarbamoyl)phenylboronic acid
4-(Isopropylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₄BNO₃
MW: 207.04 g/mol
MDL Number: MFCD04039353
CAS Number: 397843-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR