Bromotrispyrrolidinophosphonium hexafluorophosphate

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bromotripyrrolidinylphosphonium hexafluorophosphateBromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)PyBrOP Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate

COBBQA-4827-25GEA 0 GBP
COBBQA-4827-25G
Bromotrispyrrolidinophosphonium hexafluorophosphate
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Formula: C₁₂H₂₄BrF₆N₃P₂
MW: 466.2 g/mol
MDL Number: MFCD00077412
CAS Number: 132705-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR