1-Hexylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

641626-100ML 641626-800ML
SIAL641626-100MLEA 132 GBP
SIAL641626-100ML SIAL641626-800ML
1-Hexylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
1-Hexylmagnesium chloride
Formula: C₆H₁₃ClMg
MDL Number: MFCD06798087
CAS Number: 44767-62-6
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR