RSLC columns, Dionex™, SolEx™ HRP
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Dionex™ SolEx™
DION075590EA 0 GBP
DION075590 DION074400
RSLC columns, Dionex™, SolEx™ HRP
Chromatography Columns
Dionex™ SolEx™ HRP columns provide fast, easy sample preconcentration of contaminants in water by solid-phase extraction (SPE) prior to HPLC analysis.
  • Higher retention properties in both polar and hydrophobic analytes
  • No de-wetting, even with 100% aqueous phase
  • High capacity with good recoveries
  • Wide pH compatibility (0 to 14)
  • Wide range of compounds—from polar to hydrophobic analytes
Packed with a hydrophilic reversed phased (HRP) resin comprising divinylbenzene polymer with a hydrophilic bonded layer these columns are placed in the valve to automatically concentrate the analytes of interest.