Boxes with dry ice compartments, STOROpack
Supplier: Sonoco
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STOROpack
216-1337EA 374 GBP
216-1337
Boxes with dry ice compartments, STOROpack
Mailers and Shipping Containers
EPS (expanded polystyrene).
  • Lightweight, high quality material
  • Temperature assurance packaging
  • Adapted to the most sensitive products
These freezer boxes, with removable dry ice compartments, are suitable for the pharmaceutical, biotechnology and medical fields, as well as use in food service and processing industries. They offer an innovative solution for temperature-sensitive transport and storage. EPS boxes consist of 98% air and 2% polystyrene.