Ethanol 50%, Easyfix Q Path fixative reagent
Supplier: VWR Q-Path ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Ethyl alcohol
Easyfix ready to use fixative for cell samples. Pots can be used for storage and transport. Can be used as part of the Turbitec technique for cervical cancer analysis.
The fixative allows the DNA to be stored with a view to detecting HPV (human papillomavirus)
- using the Hybrid Capture II method designed by Qiagen.
- INNO-LIPA technique designed by Innogenetics.
- Papillocheck Technique designed by Greiner bio-one.
- Abbott HR HPV Technique.
Each 20 ml Pot contains 10 ml. Each 60 ml Pot contains 15 ml.
IVD registered. Instructions for use on vwr.com- just search for the product
Formula:
C₂H₅OH MW: 46.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1170 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...