3-Bromo-6-fluoro-2-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-fluoro-6-methylpyridine, 5-Bromo-2-fluoro-6-picoline

AS20817G1 AS20817G100 AS20817G25 AS20817G5
ACTIAS20817G1EA 0 GBP
ACTIAS20817G1 ACTIAS20817G100 ACTIAS20817G25 ACTIAS20817G5
3-Bromo-6-fluoro-2-methylpyridine
3-Bromo-6-fluoro-2-methylpyridine
Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03095092
CAS Number: 375368-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR