2-Nitro-1-phenylpropene

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: trans-β-Methyl-β-nitrostyrene

PROONMIAD869EA 483 GBP
PROONMIAD869
2-Nitro-1-phenylpropene
2-Nitro-1-phenylpropene
Formula: C₉H₉NO₂
MDL Number: MFCD00014720
CAS Number: 705-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR