5-Bromophthalide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromoisobenzofuran-1(3H)-one

M-Clarity™ quality level = MQ100

647187-25G 647187-5G
SIAL647187-25GEA 101 GBP
SIAL647187-25G SIAL647187-5G
5-Bromophthalide, Sigma-Aldrich®
5-Bromophthalide
Formula: C₈H₅BrO₂
MW: 213.03 g/mol
MDL Number: MFCD01797360
CAS Number: 64169-34-2
EINECS: 447-190-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR