4-Methyl-3-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51513G1 AS51513G5
ACTIAS51513G1EA 127 GBP
ACTIAS51513G1 ACTIAS51513G5
4-Methyl-3-(tributylstannyl)pyridine
4-Methyl-3-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NSn
MDL Number: MFCD11707000
CAS Number: 1204580-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR