3-Methyl-2-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51196G1 AS51196G5
ACTIAS51196G1EA 194 GBP
ACTIAS51196G1 ACTIAS51196G5
3-Methyl-2-(tributylstannyl)pyridine
3-Methyl-2-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NSn
MW: 382.16 g/mol
MDL Number: MFCD07787404
CAS Number: 259807-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR