2-Methyl-4-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51642G1 AS51642G5
ACTIAS51642G1EA 0 GBP
ACTIAS51642G1 ACTIAS51642G5
2-Methyl-4-(tributylstannyl)pyridine
2-Methyl-4-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NSn
MDL Number: MFCD16039584
CAS Number: 134914-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR