2-Fluoro-3-methyl-5-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51226G1 AS51226G5
ACTIAS51226G1EA 229 GBP
ACTIAS51226G1 ACTIAS51226G5
2-Fluoro-3-methyl-5-(tributylstannyl)pyridine
2-Fluoro-3-methyl-5-(tributylstannyl)pyridine
Formula: C₁₈H₃₂FNSn
MDL Number: MFCD09025798
CAS Number: 405556-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR