2-Fluoro-5-methyl-3-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51227G1 AS51227G5
ACTIAS51227G1EA 297 GBP
ACTIAS51227G1 ACTIAS51227G5
2-Fluoro-5-methyl-3-(tributylstannyl)pyridine
2-Fluoro-5-methyl-3-(tributylstannyl)pyridine
Formula: C₁₈H₃₂FNSn
MDL Number: MFCD09025799
CAS Number: 1025745-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR