2,6-Dibromo-4-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32132G1 AS32132G5
ACTIAS32132G1EA 32 GBP
ACTIAS32132G1 ACTIAS32132G5
2,6-Dibromo-4-nitropyridine
2,6-Dibromo-4-nitropyridine
Formula: C₅H₂Br₂N₂O₂
MW: 281.8896 g/mol
MDL Number: MFCD07369947
CAS Number: 175422-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR