Methyl-5-aminopicolinate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-aminopyridine-2-carboxylate, Methyl 5-aminopicolinate

672629-1G 672629-5G
SIAL672629-1GEA 119 GBP
SIAL672629-1G SIAL672629-5G
Methyl-5-aminopicolinate, Sigma-Aldrich®
Methyl 5-aminopicolinate
Formula: C₇H₈N₂O₂
MW: 152.1506 g/mol
MDL Number: MFCD07783939
CAS Number: 67515-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR