Ethyl-6-chloropicolinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate

PY-7261-1G PY-7261-5G
COBBPY-7261-1GEA 0 GBP
COBBPY-7261-1G COBBPY-7261-5G
Ethyl-6-chloropicolinate
Ethyl 6-chloropicolinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD09878438
CAS Number: 21190-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR