Methyl-4-chloropicolinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-chloropicolinate, Methyl 4-chloropyridine-2-carboxylate

CA-4104-25G CA-4104-5G
COBBCA-4104-25GEA 0 GBP
COBBCA-4104-25G COBBCA-4104-5G
Methyl-4-chloropicolinate
Methyl 4-chloropicolinate
Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
MDL Number: MFCD04116183
CAS Number: 24484-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR