3,5-Dibromobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
024164-1G
024164-5G
FLUO024164-1GEA
0
GBP
FLUO024164-1G
FLUO024164-5G
3,5-Dibromobenzonitrile
3,5-Dibromobenzonitrile
Formula:
C₇H₃Br₂N MW: 260.9134 g/mol |
MDL Number:
MFCD00178770 CAS Number: 97165-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...