1H,1H,5H-Octafluoropentyl acrylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate

006923-25G 006923-100G
FLUO006923-25GEA 0 GBP
FLUO006923-25G FLUO006923-100G
1H,1H,5H-Octafluoropentyl acrylate
1H,1H,5H-Octafluoropentyl acrylate
Formula: C₈H₆F₈O₂
MW: 286.12 g/mol
MDL Number: MFCD00039279
CAS Number: 376-84-1
EINECS: 206-816-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR