3,5-Dibromopyridine-2-carbonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1355-5G PY-1355-1G
COBBPY-1355-5GEA 0 GBP
COBBPY-1355-5G COBBPY-1355-1G
3,5-Dibromopyridine-2-carbonitrile
3,5-Dibromopyridine-2-carbonitrile
Formula: C₆H₂Br₂N₂
MDL Number: MFCD08062674
CAS Number: 61830-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR