3-Fluoro-5-(2,2,2-trifluoroethoxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3940-5G BB-3940-1G
COBBBB-3940-5GEA 0 GBP
COBBBB-3940-5G COBBBB-3940-1G
3-Fluoro-5-(2,2,2-trifluoroethoxy)phenylboronic acid
3-Fluoro-5-(2,2,2-trifluoroethoxy)phenylboronic acid
Formula: C₈H₇BF₄O₃
MDL Number: MFCD09475814
CAS Number: 850589-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR