2-Fluoro-4-(trifluoromethoxy)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Fluoro-4-(trifluoromethoxy)phenyl)boronic acid
FA-1878-5G
FA-1878-1G
COBBFA-1878-5GEA
0
GBP
COBBFA-1878-5G
COBBFA-1878-1G
2-Fluoro-4-(trifluoromethoxy)phenylboronic acid
2-Fluoro-4-(trifluoromethoxy)phenylboronic acid
Formula:
C₇H₅BF₄O₃ MW: 223.9200 g/mol |
MDL Number:
MFCD11040275 CAS Number: 503309-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...