3,3'-Difluorobiphenyl

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,3'-Difluoro-1,1'-biphenyl

004802-1G 004802-5G
FLUO004802-1GEA 0 GBP
FLUO004802-1G FLUO004802-5G
3,3'-Difluorobiphenyl
3,3'-Difluorobiphenyl
Formula: C₁₂H₈F₂
MW: 190.19 g/mol
MDL Number: MFCD00039216
CAS Number: 396-64-5
EINECS: 206-905-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR