3-Amino-6-chloro-4-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Amino-6-chloro-4-picoline, 5-Amino-2-chloro-4-methylpyridine, 6-Chloro-4-methylpyridin-3-amine

AS23184G1 AS23184G25 AS23184G5
ACTIAS23184G1EA 14 GBP
ACTIAS23184G1 ACTIAS23184G25 ACTIAS23184G5
3-Amino-6-chloro-4-methylpyridine
3-Amino-6-chloro-4-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD03427661
CAS Number: 66909-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR