3-Amino-5-chloro-2-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Amino-5-chloropicoline, 5-Chloro-2-methylpyridin-3-amine

AS12832G1 AS12832G5
ACTIAS12832G1EA 150 GBP
ACTIAS12832G1 ACTIAS12832G5
3-Amino-5-chloro-2-methylpyridine
3-Amino-5-chloro-2-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.5862 g/mol
MDL Number: MFCD12196974
CAS Number: 89639-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR