2-Amino-4,6-dichloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23159G1 AS23159G5
ACTIAS23159G1EA 17 GBP
ACTIAS23159G1 ACTIAS23159G5
2-Amino-4,6-dichloropyridine
2-Amino-4,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
MW: 163.0100 g/mol
MDL Number: MFCD04037218
CAS Number: 116632-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR