3-(Aminomethyl)-5-fluoropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Aminomethyl-5-fluoropyridine

PY-7306-1G PY-7306-250M
COBBPY-7306-1GEA 0 GBP
COBBPY-7306-1G COBBPY-7306-250M
3-(Aminomethyl)-5-fluoropyridine
3-(Aminomethyl)-5-fluoropyridine
Formula: C₆H₇FN₂
MW: 126.130 g/mol
MDL Number: MFCD08450424
CAS Number: 23586-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR