2-(Methylthio)-5-(tributylstannyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS51221G1
AS51221G5
ACTIAS51221G1EA
146
GBP
ACTIAS51221G1
ACTIAS51221G5
2-(Methylthio)-5-(tributylstannyl)pyridine
2-(Methylthio)-5-(tributylstannyl)pyridine
Formula:
C₁₈H₃₃NSSn |
MDL Number:
MFCD09025805 CAS Number: 611168-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...