3-Formyl-2-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methylnicotinaldehyde

AS12515G1 AS12515G5
ACTIAS12515G1EA 74 GBP
ACTIAS12515G1 ACTIAS12515G5
3-Formyl-2-picoline
3-Formyl-2-picoline
Formula: C₇H₇NO
MW: 121.14 g/mol
MDL Number: MFCD08234911
CAS Number: 60032-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR