2-Formyl-3-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Methylpyridine-2-carboxaldehyde, 3-Methylpicolinaldehyde

AS22972G1 AS22972G5
ACTIAS22972G1EA 22 GBP
ACTIAS22972G1 ACTIAS22972G5
2-Formyl-3-picoline
2-Formyl-3-picoline
Formula: C₇H₇NO
MW: 121.14 g/mol
MDL Number: MFCD02181144
CAS Number: 55589-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR