Methyl-4-hydroxy-7,8-dimethylquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6949-1G HC-6949-5G
COBBHC-6949-1GEA 0 GBP
COBBHC-6949-1G COBBHC-6949-5G
Methyl-4-hydroxy-7,8-dimethylquinoline-2-carboxylate
Methyl 4-hydroxy-7,8-dimethylquinoline-2-carboxylate
Formula: C₁₃H₁₃NO₃
MDL Number: MFCD11504931
CAS Number: 52979-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR