Methyl-4-chloro-7,8-dimethylquinoline-2-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2323-5G
HC-2323-1G
COBBHC-2323-5GEA
0
GBP
COBBHC-2323-5G
COBBHC-2323-1G
Methyl-4-chloro-7,8-dimethylquinoline-2-carboxylate
Methyl 4-chloro-7,8-dimethylquinoline-2-carboxylate
Formula:
C₁₃H₁₂ClNO₂ |
MDL Number:
MFCD11855885 CAS Number: 1133115-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...