Methyl-6-acetamido-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2511-5G HC-2511-1G
COBBHC-2511-5GEA 0 GBP
COBBHC-2511-5G COBBHC-2511-1G
Methyl-6-acetamido-4-hydroxyquinoline-2-carboxylate
Methyl 6-acetamido-4-hydroxyquinoline-2-carboxylate
Formula: C₁₃H₁₂N₂O₄
MDL Number: MFCD11855916
CAS Number: 929028-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR