Ethyl-6-fluoro-4-hydroxyquinoline-2-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6956-5G
HC-6956-1G
COBBHC-6956-5GEA
0
GBP
COBBHC-6956-5G
COBBHC-6956-1G
Ethyl-6-fluoro-4-hydroxyquinoline-2-carboxylate
Ethyl 6-fluoro-4-hydroxyquinoline-2-carboxylate
Formula:
C₁₂H₁₀FNO₃ |
MDL Number:
MFCD11504932 CAS Number: 16377-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...