Ethyl-4-hydroxyquinoline-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 4-hydroxyquinoline-2-carboxylate

AS2547G1 AS2547G5
ACTIAS2547G1EA 58 GBP
ACTIAS2547G1 ACTIAS2547G5
Ethyl-4-hydroxyquinoline-2-carboxylate
Ethyl 4-hydroxyquinoline-2-carboxylate
Formula: C₁₂H₁₁NO₃
MW: 217.2200 g/mol
MDL Number: MFCD01863732
CAS Number: 24782-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR