Methyl-7-chloro-8-fluoro-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2570-5G HC-2570-1G
COBBHC-2570-5GEA 0 GBP
COBBHC-2570-5G COBBHC-2570-1G
Methyl-7-chloro-8-fluoro-4-hydroxyquinoline-2-carboxylate
Methyl 7-chloro-8-fluoro-4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₇ClFNO₃
MDL Number: MFCD12026032
CAS Number: 1150164-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR