Methyl-8-bromo-6-fluoro-4-hydroxyquinoline-2-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6967-5G
HC-6967-1G
COBBHC-6967-5GEA
0
GBP
COBBHC-6967-5G
COBBHC-6967-1G
Methyl-8-bromo-6-fluoro-4-hydroxyquinoline-2-carboxylate
Methyl 8-bromo-6-fluoro-4-hydroxyquinoline-2-carboxylate
Formula:
C₁₁H₇BrFNO₃ |
MDL Number:
MFCD11053778 CAS Number: 442549-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...