Methyl-8-bromo-5-fluoro-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2267-5G HC-2267-1G
COBBHC-2267-5GEA 0 GBP
COBBHC-2267-5G COBBHC-2267-1G
Methyl-8-bromo-5-fluoro-4-hydroxyquinoline-2-carboxylate
Methyl 8-bromo-5-fluoro-4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₇BrFNO₃
MDL Number: MFCD11855870
CAS Number: 1133115-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR