Methyl-8-bromo-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6937-5G HC-6937-1G
COBBHC-6937-5GEA 0 GBP
COBBHC-6937-5G COBBHC-6937-1G
Methyl-8-bromo-4-hydroxyquinoline-2-carboxylate
Methyl 8-bromo-4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₈BrNO₃
MDL Number: MFCD11504929
CAS Number: 942227-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR