Methyl-6-bromo-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6944-5G HC-6944-1G
COBBHC-6944-5GEA 0 GBP
COBBHC-6944-5G COBBHC-6944-1G
Methyl-6-bromo-4-hydroxyquinoline-2-carboxylate
Methyl 6-bromo-4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₈BrNO₃
MDL Number: MFCD10001494
CAS Number: 262586-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR